Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

03-0202-00

Millipore

SMC® Human NF-L High Sensitivity Immunoassay Kit

kit sufficient for 96 wells

Synonym(s):

NF-L Single Molecule Counting kit, NF-L immunoassay kit, NF-L ultra-sensitive immunoassay, Neurofilament Light Chain kit, NfL kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.77

Quality Level

usage

 kit sufficient for 96 wells

species reactivity

human

manufacturer/tradename

SMC®

parameter

25 μL sample volume (CSF)
50 μL sample volume (serum and plasma)

assay range

intra-assay cv: ≤10%
(n=3)

sensitivity: 0.87 pg/mL
standard curve range: 0.43-1500 pg/mL
linearity: 80-120%
recovery range: 80-120%

inter-assay cv: ≤11%
(n=3)

technique(s)

single molecule counting: suitable

input

plasma (K2 EDTA)
cerebrospinal fluid (CSF)
serum

shipped in

wet ice

storage temp.

2-8°C

General description

Neurofilament Light Chain (NF-L) is a low molecular weight member of a family of proteins involved in axonal development and function. NF-L accumulation is viewed as a telltale sign of axonal damage in the brain, and is a key marker in a wide range of neurodegenerative diseases, especially as a blood-based biomarker. We are proud to aid these efforts by offering the new SMC® Human NF-L High Sensitivity Immunoassay Kit. By coupling this assay with the powerful state-of-the-art SMCxPRO® detection system, researchers can accurately assess NF-L concentrations even in low abundance in human serum, plasma, and CSF samples, enabling studies that examine the relevance of this key analyte from health to disease.

Research Category:
Neuroscience
Neurological

Research Sub-Category:
Alzheimer′s Disease
Aging
Nervous System
Neurodegenerative Diseases
Neurofilament & Neuron Metabolism
Neuronal & Glial Markers
Neuroregenerative Medicine
Neuroscience

Specificity

This assay was tested for cross-reactivity with Neurofilament Medium Chain (NF-M), Neurofilament Heavy Chain (NF-H), phospho-Tau (T181), total Tau, and Glial Fibrillary Acidic Protein (GFAP). No cross-reactivity was detected with these analytes.

Application

Used for ultrasensitivite quantification of Human NF-L in human serum, plasma, and CSF.

Features and Benefits

This SMC® kit features:
  • Accurate and reproducible performance on the state-of-the-art SMCxPRO® ultrasensitive immunoassay system.
  • Compatiblity with automated liquid handling

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service