Skip to Content
MilliporeSigma
All Photos(1)

Documents

V-046

Supelco

(±)-gamma-Tocopherol-D3 solution

500 μg/mL in methanol, certified reference material, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H45D3O2
CAS Number:
Molecular Weight:
419.70
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

500 μg/mL in methanol

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

General description

A lipid soluble form of Vitamin E, γ-Tocopherol is important to human health as an antioxidant and food additive. Vitamin E deficiency, often the result of poor nutrition or low absorption of dietary fat, is monitored in patient serum samples by LC-MS/MS or HPLC. This stable-labeled Certified Spiking Solution® is suitable for quantifying γ-tocopherol in a variety of testing applications from diagnostic testing of Vitamin E deficiency by LC-MS/MS or HPLC to quality control testing of tocopherol-based dietary supplements or pharmaceutical preparations.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service