Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S-053

Supelco

JWH-073 4-Hydroxybutyl metabolite solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21NO2
Molecular Weight:
343.42
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=CN(CCCCO)C2=C1C=CC=C2)C3=CC=CC4=C3C=CC=C4

InChI

1S/C23H21NO2/c25-15-6-5-14-24-16-21(19-11-3-4-13-22(19)24)23(26)20-12-7-9-17-8-1-2-10-18(17)20/h1-4,7-13,16,25H,5-6,14-15H2

InChI key

FIBVDFAPDNJBNI-UHFFFAOYSA-N

General description

A major urinary metabolite of JWH-073, a synthetic cannabinoid found in several blends of the herbal mixture Spice or K2. This monohydroxy analog is also known as the omega hydroxy metabolite of JWH-073.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service