Skip to Content
MilliporeSigma
All Photos(1)

Documents

P-112

Supelco

Promethazine-D3 hydrochloride solution

100 μg/mL (1% 1 M HCl in Methanol (as free base)), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17D3N2S · HCl
CAS Number:
Molecular Weight:
323.90
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (1% 1 M HCl in Methanol (as free base))

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

General description

A stable-labeled internal standard for promethazine testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, or forensic analysis. Promethazine is a phenothiazine analog prescribed for treatment of motion sickness, nausea, vomiting and allergies. Sold under the trade names Phenergan® and Phenadoz®, promethazine has antihistamine, sedative, and mild antipsychotic effects. Illicit drug users have been reported to combine promethazine with other drugs such as opioids and benzodiazepines to enhance feelings of sedation.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Phenadoz is a registered trademark of Watson Laboratories, Inc.
Phenergan is a registered trademark of Delcor Asset Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service