Skip to Content
MilliporeSigma
All Photos(1)

Documents

N-070

Supelco

Norquetiapine hydrochloride

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Norquetiapine hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N3S · HCl
CAS Number:
Molecular Weight:
331.86
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[H]Cl.C12=CC=CC=C1N=C(N3CCNCC3)C4=C(C=CC=C4)S2

InChI

1S/C17H17N3S.ClH/c1-3-7-15-13(5-1)17(20-11-9-18-10-12-20)19-14-6-2-4-8-16(14)21-15;/h1-8,18H,9-12H2;1H

InChI key

MJPARFLICDXWJT-UHFFFAOYSA-N

General description

Norquetiapine is a primary active metabolite of quetiapine, marketed as the atypical antipsychotic Seroquel® for the treatment of depression, bipolar disorder, and schizophrenia. It maintains most of the pharmacological effects of quetiapine with potencies similar to the parent drug.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Seroquel is a registered trademark of Astrazeneca UK
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service