Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N-061

Supelco

Nortilidine hydrochloride

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Nortilidine hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO2 · HCl
CAS Number:
Molecular Weight:
295.80
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CCOC(=O)[C@@]1(CCC=C[C@H]1NC)c2ccccc2

InChI

1S/C16H21NO2.ClH/c1-3-19-15(18)16(13-9-5-4-6-10-13)12-8-7-11-14(16)17-2;/h4-7,9-11,14,17H,3,8,12H2,1-2H3;1H/t14-,16+;/m1./s1

InChI key

REPHWVDUMWCCKC-XMZRARIVSA-N

General description

Nortilidine is the primary, active metabolite of tilidine, a synthetic opioid analgesic used mainly in Germany, Switzerland and Belgium for treatment of moderate to severe pain, both acute and chronic. This new Snap-N-Spike® solution standard is suitable for numerous GC/MS and LC/MS applications from clinical toxicology and forensic analysis to calibrator preparation and urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service