Skip to Content
MilliporeSigma
All Photos(1)

Documents

M-027

Supelco

(±)-MDA-D5 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-3,4-Methylenedioxyamphetamine-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D5H8NO2
CAS Number:
Molecular Weight:
184.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

NC(C([2H])([2H])[2H])([2H])C([2H])([H])C1=CC(OCO2)=C2C=C1

InChI

1S/C10H13NO2/c1-7(11)4-8-2-3-9-10(5-8)13-6-12-9/h2-3,5,7H,4,6,11H2,1H3/i1D3,4D,7D

InChI key

NGBBVGZWCFBOGO-KIUTZQKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard suitable for quantitation of MDA levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. MDA, or 3,4-methylenedioxyamphetamine, is an illicit recreational drug of the amphetamine and phenethylamine classes. Also known as tenamphetamine, MDA is abused for its psychedelic and hallucinogenic effects, similar to those of MDMA or "ecstasy".

Application


  • GC-MS analysis of amphetamines and cathinones in oral fluid: Utilizing a one-step derivatization-extraction method, this study employs (±)-MDA-D₅ as an internal standard in the rapid analysis of eleven amphetamines and cathinones by GC-MS, emphasizing its role in forensic and clinical toxicology (Mohamed, 2017).

  • Simultaneous measurement of MDMA and its metabolites in urine: A sensitive gas chromatography-mass spectrometry method incorporates (±)-MDA-D₅ to simultaneously quantify MDMA, MDEA, and their metabolites in human urine, proving crucial for pharmacokinetic and metabolic studies in clinical settings (Pirnay et al., 2006).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service