Skip to Content
MilliporeSigma
All Photos(1)

Documents

L-059

Supelco

Lorazepam-D4 glucuronide lithium salt solution

100 μg/mL (Acetonitrile:Water (50:50) (v/v), (as free acid)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14D4Cl2N2O8Li
Molecular Weight:
508.25
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Acetonitrile:Water (50:50) (v/v), (as free acid))

format

single component solution

storage temp.

-10 to -25°C

General description

This stable-labeled internal standard is suitable for quantification of lorazepam glucuronide by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications. Lorazepam glucuronide is a primary urinary metabolite of lorazepam, a benzodiazepine often prescribed for treatment of anxiety disorders.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service