Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K-006

Supelco

Ketamine-D4 hydrochloride solution

1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13D4H12ClNO · HCl
CAS Number:
Molecular Weight:
278.21
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=C([2H])C([2H])=C([2H])C([2H])=C1C2(NC)CCCCC2=O.Cl

InChI

1S/C13H16ClNO.ClH/c1-15-13(9-5-4-8-12(13)16)10-6-2-3-7-11(10)14;/h2-3,6-7,15H,4-5,8-9H2,1H3;1H/i2D,3D,6D,7D;

InChI key

VCMGMSHEPQENPE-SJSMECCZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This stable-labeled internal standard is suitable for quantitation of ketamine levels in GC/MS or LC/MS applications from clinical toxicology and forensic analysis to urine drug testing and isotope dilution methods. Ketamine, marketed under trade names Ketanest, Ketaset and Ketalar, is an anesthetic used in human and veterinary medicine. The drug is also abused at clubs, raves, or parties for its noted hallucingenic and dissociative properties and prescribed by some doctors as an off-label treatment for depression.

Application

This stable-labeled internal standard is suitable for quantitation of ketamine levels in GC/MS or LC/MS applications from clinical toxicology and forensic analysis to urine drug testing and isotope dilution methods. Ketamine, marketed under trade names Ketanest®, Ketaset® and Ketalar®, is an anesthetic used in human and veterinary medicine. The drug is also abused at clubs, raves, or parties for its noted hallucingenic and dissociative properties and prescribed by some doctors as an off-label treatment for depression.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Ketalar is a registered trademark of Par Sterile Products, LLC
Ketanest is a registered trademark of Pfizer, Inc.
Ketaset is a registered trademark of Zoetis W LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service