Skip to Content
MilliporeSigma
All Photos(1)

Documents

G-013

Supelco

Ginkgo Biloba Terpene Lactones Mix

100 μg/mL each component in acetonitrile, ampule of 1.0 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O10
Molecular Weight:
424.40
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Bilobalid
Ginkgolide A
Ginkgolide B
Ginkgolide C
Ginkgolide J

packaging

ampule of 1.0 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL each component in acetonitrile

application(s)

food and beverages

format

multi-component solution

storage temp.

−20°C

General description

Discover the entire range of phytochemical certified reference material solutions from Cerilliant


Ginkgo biloba extract is a mixture of biologically active secondary metabolites. The main constituents are bilobalide (BB), ginkgolide A (GA), ginkgolide B (GB) and ginkgolide C (GC). Ginkgo biloba extracts include around 300 compounds, with major constituents being flavonoids and terpene lactones. Terpene lactones make up to 6% of the total extract.

Biochem/physiol Actions

Ginkgo biloba terpene lactone constitutes a major part of Ginkgo Biloba extract (GBE), which is used as a phytomedicine, and is used for multiple disorders including CNS (central nervous system), circulatory, cardiovascular, respiratory and renal diseases. Extracts from the leaves of Gingko plants are thought to have anti-stress, anti-cancer, neuroprotective, cardioprotective and memory enhancing properties. Terpenoid lactones are thought play a complementary role in these effects of GBE.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yong Joo Park et al.
Asian Pacific journal of cancer prevention : APJCP, 16(15), 6317-6325 (2015-10-06)
Ginkgo biloba extract (GBE) is a popular phytomedicine and has been used for disorders of the central nervous system, cardiovascular, renal, respiratory, and circulatory diseases. Although GBE is a complex mixture of over 300 compounds, its major components are 24%
S Mahadevan et al.
Journal of food science, 73(1), R14-R19 (2008-01-24)
The new age of nutraceuticals is now embracing the centuries old herbal extract of Ginkgo biloba (Mantissa Plantarum Altera, 1771, Ginkgoceae). The standardized preparation of the Ginkgo leaf extract (EGb 761) contained 2 main bioactive constituents, flavonoid glycosides (24%) and
Mostafa K Mesbah et al.
Farmaco (Societa chimica italiana : 1989), 60(6-7), 583-590 (2005-06-07)
The biologically active secondary metabolites of Ginkgo biloba extract EGb 761 in phytopharmaceuticals were analyzed using two simple, rapid, accurate and sensitive HPLC methods. The proposed methods were successfully applied in the determination of terpenes and flavonoids in four phytopharmaceutical
Ping Huang et al.
Journal of pharmaceutical and biomedical analysis, 100, 138-144 (2014-08-29)
The ginkgo terpene lactones (GTL), mainly including bilobalide (BB), ginkgolide A (GA), ginkgolide B (GB) and ginkgolide C (GC) possess different biological activities such as peripheral vasoregulation, platelet-activating factor (PAF) receptor antagonism, neuroprotective properties and prevention of membrane damage caused

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service