Skip to Content
MilliporeSigma
All Photos(1)

Documents

D-120

Supelco

Docosapentaenoic acid (cis-7,10,13,16,19)

500 μg/mL in ethanol, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O2
CAS Number:
Molecular Weight:
330.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

500 μg/mL in ethanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCCCC(O)=O

InChI

1S/C22H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h3-4,6-7,9-10,12-13,15-16H,2,5,8,11,14,17-21H2,1H3,(H,23,24)/b4-3-,7-6-,10-9-,13-12-,16-15-

InChI key

YUFFSWGQGVEMMI-JLNKQSITSA-N

General description

A Certified Spiking Solution® suitable for use in mass spectrometry-based fatty acid testing applications such as assessment of cardiovascular disease risk and fatty acid deficiency, and detection and quantification of docosapentaenoic acid in nutraceuticals and dietary supplements. Docosapentaenoic acid (cis-7,10,13,16,19), also known as DPA, is an omega-3 fatty acid associated with neurological and ocular health benefits. Studies suggests that DPA as well as other fatty acids can serve as biomarkers for cardiovascular disease, and nutritional and metabolic disorders.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service