Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-241

Supelco

Cannabicitran (CBT) solution

1 mg/mL (Degassed Acetonitrile), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (Degassed Acetonitrile)

format

single component solution

storage temp.

-10 to -25°C

InChI

1S/C21H30O2/c1-5-6-7-8-14-11-17-19-15-13-21(4,23-18(19)12-14)10-9-16(15)20(2,3)22-17/h11-12,15-16H,5-10,13H2,1-4H3/t15-,16+,21+/m0/s1

InChI key

IXJXRDCCQRZSDV-GCKMJXCFSA-N

General description

Cannabicitran, also known as CBT, is a minor phytocannabinoid. This certified Snap-N-Spike® solution is suitable as starting material in preparation of calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis, clinical toxicology, pharmaceutical research to cannabis testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

An accurate and precise HPLC-PDA measurement of 32 cannabinoids and 2 cannflavins in hemp samples using a single extraction in less than 32 minutes.

An accurate and precise HPLC-PDA measurement of 32 cannabinoids and 2 cannflavins in hemp samples using a single extraction in less than 32 minutes.

An accurate and precise HPLC-PDA measurement of 32 cannabinoids and 2 cannflavins in hemp samples using a single extraction in less than 32 minutes.

An accurate and precise HPLC-PDA measurement of 32 cannabinoids and 2 cannflavins in hemp samples using a single extraction in less than 32 minutes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service