Skip to Content
MilliporeSigma
All Photos(1)

Documents

C-219S

Supelco

Cannabinoid Mixture (Neutrals) - 8 Component solution

500 μg/mL in methanol ((each analyte)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O2
Molecular Weight:
286.41
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Cannabidiol, 500 μg/mL
Cannabigerol (CBG), 500 μg/mL
Cannabinol, 500 μg/mL
CBC (Cannabichromene), 500 μg/mL
CBDV (Cannabidivarin), 500 μg/mL
(-)-delta8-THC, 500 μg/mL
(-)-delta9-THC, 500 μg/mL
THCV, 500 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

500 μg/mL in methanol ((each analyte))

format

multi-component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application


  • Specialized solid phase extraction for cannabinoid analysis: Research demonstrated the design and application of a specialized solid phase extraction column tailored for the analysis of three cannabinoid compounds in hemp. This study underscores the importance of precise analytical tools in cannabinoid profiling, offering significant implications for pharmaceutical research and biochemistry applications (Shen et al., 2021).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service