Skip to Content
MilliporeSigma
All Photos(1)

Documents

C-169

Supelco

4-Chloroethcathinone hydrochloride solution

1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14ClNO · HCl
CAS Number:
Molecular Weight:
248.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC=C(C(C(NCC)C)=O)C=C1.Cl

InChI

1S/C11H14ClNO/c1-3-13-8(2)11(14)9-4-6-10(12)7-5-9/h4-8,13H,3H2,1-2H3

InChI key

RNBCGEIZCHBTGL-UHFFFAOYSA-N

General description

4-Chloroethcathinone, also called 4-CEC, is a stimulant and designer drug found in "bath salts" sold at head shops and over the Internet. These synthetic cathinones offer recreational uses that mimic the effects of illegal drugs such as cocaine, methamphetamine, and LSD. This Certified Spiking Solution® is suitable as starting material in calibrators and controls for GC/MS or LC/MS bath salt testing applications in forensic analysis, urine drug testing or clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik Postma et al.
Oecologia, 174(3), 631-638 (2013-10-16)
Egg components are important mediators of prenatal maternal effects in birds and other oviparous species. Because different egg components can have opposite effects on offspring phenotype, selection is expected to favour their mutual adjustment, resulting in a significant covariation between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service