Key Documents
Z748242
Sigma-Aldrich
Load flask for Aldrich® Kugelrohr apparatus
capacity 500 mL, joint: ST/NS 14/20
Synonym(s):
distillation apparatus, distilling apparatus, fractional distillation apparatus, glass distillation apparatus, kugelrohr apparatus, kugelrohr distillation, short path distillation apparatus, vacuum distillation apparatus
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
glass
single-neck
spherical
Joint
joint: ST/NS14/20
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Sigma-Aldrich
capacity
500 mL
General description
Heavy wall glass flask with solid support that attaches to Kugelrohr motor drive coupling. Fill load flask one-third full with distillable material.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC