Key Documents
Z742605
myVolt Touch power supply
AC/DC input 230 V AC, EuroPlug
About This Item
UNSPSC Code:
41105300
NACRES:
JA.13
AC/DC input
5 - 400 mA (Max Wattage: 80W; Voltage Output: 10-300V (5V increments))
feature
output terminals: 4 pairs
touchscreen control; Dual orientation, auto rotating display
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific E2301-E
compatibility
with most brands of gel boxes
General description
The Accuris myVolt Touch is a dependable electrophoresis power supply that includes all of the performance and specifications needed for efficient seperations of nucleic acids and proteins. Perfect for agarose gels and protein seperations, the myVolt Touch incorporates some very unique user-friendly features: Touch-screen Control Panel: Setting up an electrophoresis run with the myVolt Touch couldn′t be easier... simply touch the parameter that you want to change and then touch "start" to begin the run.
Two sets of output jacks are available to connect up to 2 gel rigs and the instruments can securely be stacked for use with additional gel rigs.
Legal Information
myVolt is a trademark of Benchmark Scientific, Inc.