Key Documents
Z691577
Mini Bin aluminum foil
W 6 in. (153 mm)
Synonym(s):
aluminum foil, tube foil, tube sealing foil
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
material
aluminum film
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS23536A
W × L × thickness
15.3 cm × 152.40 m -0.015 mm
W
6 in. (153 mm)
General description
Designed for easy use and minimal waste. Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes and petri dishes. Ideal for autoclaving, freezing, incubating, protecting lab ware, lining, sealing and general storage.
Two sizes available. Designed exclusively for the Mini Bin Foil Dispenser.
Legal Information
Mini Bin is a trademark of Heathrow Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Articles
A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.
A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.
A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.
A complete workflow for estimating the total polyphenol content of apple and banana samples, with gallic acid standard addition curve protocol.