Key Documents
Z689963
Sigma-Aldrich
Aldrich® column clamp
size L
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
size
L
L × diam.
211 mm × 11 mm , arm
jaw capacity
64-140 mm
General description
Corrosion resistant stainless steel. Ideal for holding chromatography columns and large cylindrical glassware. A large, flat thumbscrew/worm drive permits maximum tightening or removal in minimum time.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC