Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z678686

2mag bioMIXdrive 1 magnetic stirrer

for use with bioMIXcontrol S control unit

Synonym(s):

induction stirrer, magnetic induction stir plate, magnetic wear-free stirrer, stirrer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103806
NACRES:
NB.72

material

stainless steel plate

feature

CE compliant
positions 1

packaging

pack of 1 ea

manufacturer/tradename

2mag 80001

parameter

5-250 rpm speed

plate size × W × L

× 180 mm × 180 mm

stirrer volume

5-5000 mL , per stirring position

Looking for similar products? Visit Product Comparison Guide

General description

Warming- and maintenance-free magnetic stirrer with large surface, up to 5,000 mL stirring volume, also suitable for viscous culture media.

Maintenance-free 2mag-stepper-motor-drive for extremely slow and 100% jerk-free and gentle mixing of cell cultures, extended speed range from 5 up to 250 rpm, jerk-free, even and smooth stirring also at slow speeds, suitable for culture flasks with spinning propeller- and spinning ball systems.

Fully encapsulated and hermetically closed stainless steel housing, IP68, water-, dust- and germ-proof, extremely robust construction and performance, easy to clean and maximum firm, submersible in water baths, designed for the warming-free long term operation in CO2-incubators, individual measurements on request.

2mag bioMIXcontrol MS4 unit is needed and are sold separately.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service