Key Documents
Z678023
Finnpipette® F2 multichannel pipette
volume 1-10 μL, 12 channel
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
12-channel
color-coded pink
electronic: no
packaging
pack of 1 ea
manufacturer/tradename
Finn 4662040
volume
1-10 μL
General description
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Double action pipetting button with very light and smooth pipetting action
  • Fully autoclavable
  • Super blow-out piston with volumes of 50 μl and below
Compatibility
10, Flex 10
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries