Key Documents
Z675997
RF1000 pipette filler
a lightweight manual pipette filler uniquely designed for one-handed operation
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
plastic body
silicone bulb
manufacturer/tradename
Heathrow Scientific HS1000RFU
size
180 mm × 45 mm × 120 mm
volume
1-100 mL
General description
Heathrow Scientifics RF1000 is a lightweight manual pipette filler uniquely designed for one-handed operation. Simply squeeze the main bulb, and push the toggle forward to aspirate or back to dispense. Push the blowout bulb to expel any liquid remaining in the pipette.
The RF1000 accommodates graduated or volumetric pipettes (glass or plastic) and can be used with volumes ranging from 1 mL to 100 mL. Includes a hydrophobic 0.45 μm filter and a magnetic holder for metal surface mount or wall mount. The polypropylene nozzle is autoclavable.
Legal Information
RF1000 is a trademark of Heathrow Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)