Key Documents
Z675482
Sterile economy sample spoon
20 mL
Synonym(s):
Economy Sample Spoon, Sterile Sample Spoon, dispensing tool
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
description
PS
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art F36740-00005
General description
Moulded of lightweight and rigid, white high impact polystyrene. Individually packaged sterile spoons for one time use packed 100 per box.
  • Use to collect specific volumes of dry or liquid materials
  • Remains upright when filled or empty for use as a weighing boat