Key Documents
Z657174
IKA® shaker attachment for HS/KS 260 basic and control shakers
AS 260.5 seperating funnel attachment, 1/cs
Synonym(s):
benchtop shakers, lab shakers, laboratory shakers
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 3120900
suitability
suitable for (for shaking out, salting out, extracting, eluting and sample enrichment)
General description
IKA® AS 260.5 Seperating Funnel Attachment
  • For shaking out, salting out, extracting, eluting, enriching.
  • The 3 clamping rolls (included in delivery) are height-adjustable for adaption to different separating funnel sizes.
  • The separating funnels are secured with O-rings (5 O-rings included).
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG