Key Documents
Z548979
Sigma-Aldrich
Aldrich® splash protector
female joint: ST/NS 29/32, male joint: ST/NS 14/20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
female joint: ST/NS29/32
male joint: ST/NS14/20
General description
Effectively prevents upward movement of foaming or bumping liquids in rotary evaporator traps. Unique design has two baffles with horizontally opposing holes that may also be used with other distillation techniques.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC