Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z509043

Sigma-Aldrich

Pipette bulb

volume 3 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

material

natural rubber
thimble blue

packaging

pkg of 24 ea

manufacturer/tradename

200-3A

bulb I.D.

7 mm

volume

3 mL

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service