Key Documents
Z409480
Sigma-Aldrich
Weighing funnel
size M, polypropylene
Synonym(s):
Weighing Funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.71
material
polypropylene
size
M
packaging
pkg of 50 ea
General description
  • Sit flat on balance - No rolling, no spilling!
  • Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer
  • Residual sample can be rinsed to guarantee 100% transfer
  • Suitable for safe handling of hazardous materials
  • Available in five different sizes: extra small, small, medium, large, and extra large