Key Documents
Z278769
Sigma-Aldrich
Bytac® surface protection laminate
PTFE, size 25 in. × 15 ft
Synonym(s):
bench protection, benchtop laminate, worksurface protection
About This Item
UNSPSC Code:
41122300
NACRES:
NB.35
material
PTFE
vinyl backing
packaging
pack of 1 ea
size
25 in. × 15 ft
Related Categories
General description
Add the benefits of FEP to any flat surface. Bytac protects surfaces from absorption of toxic and radioactive materials. The laminate consists of FEP resin supported on a backing of vinyl or aluminum. The backing has a pressure sensitive acrylic adhesive that adheres to most surfaces. Vinyl backing is recommended for applications up to 200 °F (93 °C). Aluminum backing is for high-temperature applications and can be used up to 360 °F (182 °C).
  • non-stick; easy to clean
  • chemically inert
Legal Information
Bytac is a registered trademark of Saint-Gobain Performance Plastics Corp.