Documents
Z236756
Sigma-Aldrich
WypAll® wipers
L × W 9 3/4 in. × 16 3/4 in.
Synonym(s):
cleaning tissue
About This Item
UNSPSC Code:
41103200
NACRES:
NB.35
packaging
pkg of 126 wipes
manufacturer/tradename
Kimberly Clark 34790
L × W
9 3/4 in. × 16 3/4 in.
General description
Strong-even when wet. 4-ply nylon-reinforced embossed tissues are made from 100% recycled paper. Convenient dispensing systems reduce waste, protect wipers, and prevent pilferage.
Legal Information
WypAll is a registered trademark of Kimberly-Clark Worldwide, Inc.
Visit the Document Library
Customers Also Viewed
Slide 1 of 2
1 of 2