Key Documents
Z173363
Sigma-Aldrich
Recrystallizer
male Joints: ST/NS 14/20 (4), porosity 25-50 μm
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
label
DD
Joint
male Joints: ST/NS14/20 (4)
pore size
25-50 μm porosity
Related Categories
General description
Double-tube.
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.