Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WIMWG40007

Wilmad® NMR tubes 10 mm diam., economy

cap: no, high throughput, L 7 in.

Synonym(s):

Wilmad nmr tubes, nmr sample tubes, nmr tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121705
NACRES:
SB.32

material

borosilicate glass tube
colorless tube

feature

cap: no high throughput

packaging

pack of 100 ea

manufacturer/tradename

Wilmad WG-4000-7

parameter

600 MHz frequency

L

7 in.

O.D.

9.994 mm

camber

60 μm

tube diam.

10 mm

wall thickness

0.60 mm

General description

10 mm Economy NMR Tubes are manufactured from borosilicate glass that meets the ASTM Type 1 Class B standard and recognized as N51A. Due to its nature, this glass has much less thermal-shock resistance than Class A glass used for our Precision Tubes. It may contain Fe2O3 which is paramagnetic around 1200 ppm and has certain impact on shimming quality at high field over 600 MHz.

10 mm NMR tubes have an averaged camber of 60 micron to guarantee the spectrum resolution for small molecules (MW<500) samples up to 600 MHz.

Legal Information

Wilmad is a registered trademark of SP Industries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service