Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WHA3454651

Whatman® cellulose chromatography papers

54 SFC Roll, W × L 1.5 in. × 300 ft

Synonym(s):

Whatman paper, Z742503, blotting papers, cellulose membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115727
NACRES:
NB.24

material

cellulose
cellulose membrane
roll

grade

for chromatography

sterility

non-sterile

packaging

pkg of 1 ea

manufacturer/tradename

Whatman 3454-651
Whatman Article No. 28419385 (US reference)

parameter

1.8 cm/hr speed
180mm/30 min flow rate

W × L

1.5 in. × 300 ft

Looking for similar products? Visit Product Comparison Guide

General description

54 SFC cellulose chromatography roll is a 0.18 hardened paper for routine chromatography. Thin (0.18 mm) hardened paper with high speed (180 mm/30 min.) and fair to good resolution. Recommended for routine chromatography. High wet strength.

Features and Benefits

  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana Campuzano et al.
Applied and environmental microbiology, 75(18), 5972-5980 (2009-07-28)
A Streptococcus mitis genomic DNA fragment carrying the SMT1224 gene encoding a putative beta-galactosidase was identified, cloned, and expressed in Escherichia coli. This gene encodes a protein 2,411 amino acids long with a predicted molecular mass of 268 kDa. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service