Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WHA2300731

Whatman® Benchkote® surface protector

Benchkote surface protector, reel, 460 mm x 50 m, 1/pk

Synonym(s):

Z671479, absorbent papers, bench protectors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122300
NACRES:
NB.35

packaging

pack of 1

manufacturer/tradename

Whatman 2300-731
Whatman Article No. 28418848 (US reference)

size

46 cm × 50 m

Looking for similar products? Visit Product Comparison Guide

General description

Benchkote

Benchkote® is an absorbent, impermeable material designed to protect laboratory surfaces against hazardous spills. The material features a high-quality, smooth, absorbent Whatman paper which quickly absorbs liquid spills and a laminated polyethylene layer that prevent flow through to the working surface. After use the sheet is incinerated or disposed of acording to local regulations.

Benchkote Plus

Benchkote Plus is a thicker, more absorbent material for more demanding applications and can absorb in excess of 0.75 liters of water per square meter.

Application

Applications:
  • Containing radiochemical spillage and avoiding contamination
  • Recovering spillage of expensive materials
  • Protecting hard surfaces to lessen impact
  • Water or solvent wick for humidity chambers
  • Lining of chemical cabinets, laboratory bench drawers, and laboratory hoods

Features and Benefits

Features and Benefits:
  • Material is very strong, making it tear resistant, wet or dry
  • Smooth white surface can be written on with ink or pencil and lies flat
  • Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Use polyethylene side up to collect deposits without absorption
  • Paper side quickly absorbs liquid spills preventing liquids from going through to the work surface
  • Spillages are trapped in the absorbent paper
  • Benchkote can be incinerated after use; the polyethylene layer does not melt or drip but is rapidly consumed in the flames

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Benchkote is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service