Key Documents
WHA2017013
Whatman® Antibiotic Assay Discs
diam. 13 mm
Synonym(s):
Whatman paper, Z741337
About This Item
UNSPSC Code:
41111700
NACRES:
NB.79
packaging
pkg of 1000 ea
manufacturer/tradename
Whatman 2017-013
Whatman Article No. 28418662 (US reference)
diam.
13 mm
General description
For determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
Features and Benefits
  • The antibiogram allows rational and selective chemotherapy.
  • Test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated.
  • The size of the inhibition zone is a measure for the effectiveness of the substances.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva