Skip to Content
MilliporeSigma
All Photos(1)

Documents

W6985

Octanoic acid-1-13C

API for Clinical Studies

Synonym(s):

Caprylic acid-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)613CO2H
CAS Number:
Molecular Weight:
145.20
MDL number:
UNSPSC Code:
12352106

grade

cGMP

isotopic purity

≥99 atom % 13C

refractive index

n20/D 1.4284 (lit.)

bp

237 °C (lit.)

mp

16 °C (lit.)

mass shift

M+1

storage temp.

15-25°C

SMILES string

CCCCCCC[13C](O)=O

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)/i8+1

InChI key

WWZKQHOCKIZLMA-VJJZLTLGSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service