Skip to Content
MilliporeSigma
All Photos(1)

Documents

W520802

Sigma-Aldrich

1,4-Butanedithiol

≥97%, FG

Synonym(s):

1,4-Dimercaptobutane, Tetramethylene dimercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)4SH
CAS Number:
Molecular Weight:
122.25
Beilstein:
1697080
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.103
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.529 (lit.)

bp

105-106 °C/30 mmHg (lit.)

density

1.042 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; sulfurous

SMILES string

[K+].[K+].[O-]S([O-])=O

InChI

1S/C4H10S2/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

SMTOKHQOVJRXLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Butanedithiol, an alkanedithiol, is a food odorant. Its odor threshold is similar to its other homologs (propane-1,3-dithiol and pentane-1,5-dithiol).

Application


  • Hydroxyapatite nanoparticle-modified porous bone grafts with improved cell attachment.: This study demonstrates the application of 1,4-butanedithiol in modifying hydroxyapatite nanoparticles to enhance cell attachment on porous bone grafts, presenting significant implications for biomedical engineering and regenerative medicine (Dhavalikar et al., 2023).

  • Engineering Toolbox for Systematic Design of PolyHIPE Architecture.: This paper explores the role of 1,4-butanedithiol in the systematic design of polymer architectures, offering insights into materials science that could influence various industrial applications (Dhavalikar et al., 2021).

  • Thiol-ene miniemulsion polymerization of a biobased monomer for biomedical applications.: The study utilizes 1,4-butanedithiol in thiol-ene miniemulsion polymerization, aiming at creating biocompatible polymers for biomedical applications, thus advancing the field of biomaterials (Machado et al., 2017).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure-Odor Correlations in Homologous Series of Alkanethiols and Attempts To Predict Odor Thresholds by 3D-QSAR Studies
Polster J and Schieberle P
Journal of Agricultural and Food Chemistry, 63(5), 1419-1432 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service