Skip to Content
MilliporeSigma
All Photos(1)

Documents

W402633

Sigma-Aldrich

Benzyl hexanoate

≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
FEMA Number:
4026
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.316
NACRES:
NA.21

grade

FG

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥97%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apricot; gardenia; green; jasmine; fruity; pineapple

InChI

1S/C13H18O2/c1-2-3-5-10-13(14)15-11-12-8-6-4-7-9-12/h4,6-9H,2-3,5,10-11H2,1H3

InChI key

HRSXWUSONDBHSP-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service