Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W363308

Sigma-Aldrich

cis-3-Hexenyl phenylacetate

≥98%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
218.29
FEMA Number:
3633
EC Number:
Council of Europe no.:
10682
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.805
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

contains

α-Tocopherol, synthetic as stabilizer

refractive index

n20/D 1.502 (lit.)

bp

292-297 °C (lit.)

density

0.992 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; honey; floral

SMILES string

[H]\C(CC)=C(/[H])CCOC(=O)Cc1ccccc1

InChI

1S/C14H18O2/c1-2-3-4-8-11-16-14(15)12-13-9-6-5-7-10-13/h3-7,9-10H,2,8,11-12H2,1H3/b4-3-

InChI key

FJKFIIYSBXHBCT-ARJAWSKDSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service