Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W316008

Sigma-Aldrich

Furfuryl methyl sulfide

≥97%, FG

Synonym(s):

2-(methylsulfanylmethyl)furan, Methyl furfuryl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8OS
CAS Number:
Molecular Weight:
128.19
FEMA Number:
3160
EC Number:
Council of Europe no.:
11482
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.053
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA

Assay

≥97%

refractive index

n20/D 1.521 (lit.)

bp

64-65 °C/15 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

garlic; onion; pungent; alliaceous; sulfurous

SMILES string

CSCc1ccco1

InChI

1S/C6H8OS/c1-8-5-6-3-2-4-7-6/h2-4H,5H2,1H3

InChI key

SKSFHXVDHVKIBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • RIFM fragrance ingredient safety assessment, furfuryl methyl sulfide, CAS Registry Number 1438-91-1.: This paper presents a safety assessment of furfuryl methyl sulfide, identifying it as a key component in fragrances and exploring its toxicological profile and safe use levels (Api et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wentian Chen et al.
Journal of food science and technology, 56(2), 937-950 (2019-03-25)
Anthocyanin-rich concentrates from different red radish can be used as natural food colorants. However, the development of off-flavor during extraction has been major challenge in processing industries. This work aimed to evaluate the effect of sodium chloride (NaCl) concentration in
Xuebo Song et al.
Food chemistry, 297, 124959-124959 (2019-06-30)
Volatile sulfur-containing compounds (VSCs) often exist at extremely low concentrations, making them difficult to be determined. The VSCs in Laobaigan (LBG) Baijiu were analyzed by headspace solid-phase microextraction (HS-SPME) coupled with GC × GC-SCD, by which 12 VSCs were identified. Among the 65

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service