Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W292516

Sigma-Aldrich

Propyl acetate

natural, ≥97%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2CH3
CAS Number:
Molecular Weight:
102.13
FEMA Number:
2925
Beilstein:
1740764
EC Number:
Council of Europe no.:
192c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.002
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

3.5 (vs air)

vapor pressure

25 mmHg ( 20 °C)

Assay

≥97%

form

liquid

autoignition temp.

842 °F

expl. lim.

1.7 %, 37 °F
8 %

refractive index

n20/D 1.384 (lit.)

bp

102 °C (lit.)

mp

−95 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

celery; fruity; pear

SMILES string

CCCOC(C)=O

InChI

1S/C5H10O2/c1-3-4-7-5(2)6/h3-4H2,1-2H3

InChI key

YKYONYBAUNKHLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.2 °F - closed cup

Flash Point(C)

11.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Gustavsson et al.
Bioorganic & medicinal chemistry, 5(12), 2173-2183 (1998-02-12)
The enantiomers of cis- and trans-3-(4-propyl-cyclopent-2-enyl) propyl acetate, which are conformationally constrained analogues of (Z)-5-decenyl acetate (1), a sex pheromone component of the turnip moth, Agrotis segetum, have been synthesized and tested using the electrophysiological single-sensillum technique. The analogues mimic
Lan-Chun Nie et al.
Zhi wu sheng li yu fen zi sheng wu xue xue bao = Journal of plant physiology and molecular biology, 31(6), 663-667 (2005-12-20)
Aroma volatiles from apple (Malus domestica Borkh. var. Starkrimson) fruit at different stages of maturity were collected by solid adsorbent-Tenax-GC and determined by thermodesorption and GC-MS. Production of propyl acetate, butyl acetate, ethyl 2-methyl-butanoate and total ester volatiles and changes
[Gas chromatographic determination of propyl acetate in air].
N N Sirotkina et al.
Gigiena i sanitariia, (7)(7), 46-47 (1981-07-01)
S Puglisi-Allegra et al.
Pharmacology, biochemistry, and behavior, 13(2), 287-290 (1980-08-01)
An automated technique for measuring simultaneously aggressive behavior and motor activity in mice is described. This technique allows measure of aggressive behavior and motor activity continuously throughout long-lasting periods without disturbing the animals except for feeding, watering and cleaning the
C A Long et al.
Radiology, 180(1), 47-49 (1991-07-01)
The in vitro effectiveness of three new cholesterol gallstone solvents that have a potential for clinical use--ethyl propionate, isopropyl acetate, and n-propyl acetate--was compared with that of the well-known cholesterol solvent methyl tert-butyl ether (MTBE). Ten sets of matched gallstones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service