Skip to Content
MilliporeSigma
All Photos(1)

Documents

W258318

Sigma-Aldrich

Hydroxycitronellal

≥95%, FCC, FG

Synonym(s):

7-Hydroxy-3,7-dimethyl-octanal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
FEMA Number:
2583
EC Number:
Council of Europe no.:
100
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.012
NACRES:
NA.21

biological source

synthetic

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

composition

contains IFRA and EU 1223/2009 restricted Hydroxycitronellal

refractive index

n20/D 1.448 (lit.)

bp

257 °C (lit.)

density

0.923 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

hydroxycitronellal

Organoleptic

green; melon; floral; sweet

SMILES string

[H]C(=O)CC(C)CCCC(C)(C)O

InChI

1S/C10H20O2/c1-9(6-8-11)5-4-7-10(2,3)12/h8-9,12H,4-7H2,1-3H3

InChI key

WPFVBOQKRVRMJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxycitronellal is an odorant generally used in cosmetics, washing- and cleaning agents and also as a flavoring agent for foods.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A validated UPLC?MS/MS method for biomonitoring the exposure to the fragrance 7-hydroxycitronellal.
Stoeckelhuber M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1068, 261-267 (2017)
M Cluzel-Tailhardat et al.
Toxicology letters, 174(1-3), 98-109 (2007-10-16)
A critical step in the induction of allergic contact dermatitis is the interaction of haptens with immature dendritic cells (iDC) leading to their activation. Therefore iDC appear as suitable targets for the evaluation of the sensitizing properties of haptens with
B Kreilgård et al.
Journal of the American Academy of Dermatology, 21(4 Pt 2), 836-838 (1989-10-01)
The choice of vehicle for patch test materials is important for the bioavailability and stability of the allergens. Examples are given from the standardization of the TRUE Test and include caine mix, Kathon CG, and fragrance mix.
Clarissa D Villarama et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 42(11), 1719-1725 (2004-09-08)
The clinical relevance of patch test reactions is often difficult to determine. Use tests have been developed to further evaluate the significance of patch test results. We review studies that have defined correlations between the threshold concentrations at patch testing
Susan P Felter et al.
Regulatory toxicology and pharmacology : RTP, 37(1), 1-10 (2003-03-29)
The National Academy of Science (NAS) risk assessment paradigm has been widely accepted as a framework for estimating risk from exposure to environmental chemicals (NAS, 1983). Within this framework, quantitative risk assessments (QRAs) serve as the cornerstone of health-based exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service