Skip to Content
MilliporeSigma
All Photos(1)

Documents

W255602

Sigma-Aldrich

γ-Hexalactone

≥98%, FCC, FG

Synonym(s):

γ-Caprolactone, γ-Ethyl-γ-butyrolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
FEMA Number:
2556
Beilstein:
6114822
EC Number:
Council of Europe no.:
2254
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.021
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.439 (lit.)

bp

219 °C (lit.)

density

1.023 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coconut; coumarin; creamy; sweet

SMILES string

CCC1CCC(=O)O1

InChI

1S/C6H10O2/c1-2-5-3-4-6(7)8-5/h5H,2-4H2,1H3

InChI key

JBFHTYHTHYHCDJ-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • RIFM fragrance ingredient safety assessment, γ-hexalactone, CAS Registry Number 695-06-7.: This article provides a comprehensive safety assessment of γ-Hexalactone, a key fragrance ingredient, supporting its safe use under current practices and concentration levels in consumer products (Api et al., 2022).

  • Production of Hydroxy Fatty Acids, Precursors of γ-Hexalactone, Contributes to the Characteristic Sweet Aroma of Beef.: This study highlights the biochemical pathways involved in the formation of γ-Hexalactone through the production of hydroxy fatty acids, significantly impacting the flavor profile of beef (Ueda et al., 2022).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Khersonsky et al.
Biochemistry, 44(16), 6371-6382 (2005-04-20)
PON1 is the best-studied member of a family of enzymes called serum paraoxonases, or PONs, identified in mammals (including humans) and other vertebrates as well as in invertebrates. PONs exhibit a range of important activities, including drug metabolism and detoxification
H Thoma et al.
Journal of chromatography, 309(1), 17-32 (1984-07-13)
A two-step ultrafiltration method combined with anion-exchange chromatography is described for the separation of lipophilic and strongly lipophobic acids occurring in human blood, serum and plasma. After treatment with diazomethane, the acid fractions are separated further by gas chromatography. The
Profile of volatile metabolites in human urine.
A Zlatkis et al.
Clinical chemistry, 17(7), 592-594 (1971-07-01)
Laura E Korhonen et al.
Journal of medicinal chemistry, 48(11), 3808-3815 (2005-05-27)
The purpose of this study was to determine the cytochrome P450 1A2 (CYP1A2) inhibition potencies of structurally diverse compounds to create a comprehensive three-dimensional quantitative structure-activity relationship (3D-QSAR) model of CYP1A2 inhibitors and to use this model to predict the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service