Skip to Content
MilliporeSigma
All Photos(1)

Documents

W234907

Sigma-Aldrich

Cyclohexyl acetate

≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC6H11
CAS Number:
Molecular Weight:
142.20
FEMA Number:
2349
EC Number:
Council of Europe no.:
217
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.027
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.439 (lit.)

bp

172-173 °C (lit.)

density

0.966 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

musty; fruity; sweet

SMILES string

CC(=O)OC1CCCCC1

InChI

1S/C8H14O2/c1-7(9)10-8-5-3-2-4-6-8/h8H,2-6H2,1H3

InChI key

YYLLIJHXUHJATK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexyl acetate is a monocyclic ester that can be used as a flavoring agent and fragrance ingredient.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

136.4 °F - DIN 51758

Flash Point(C)

58 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragrance material review on cyclohexyl acetate
Bhatia SP, et al.
Food And Chemical Toxicology, 46(12), S52-S55 (2008)
A toxicologic and dermatologic assessment of cyclic acetates when used as fragrance ingredients
Belsito D, et al.
Food And Chemical Toxicology, 46(12), S1-S27 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service