Key Documents
SYNP140010M
Synthware peptide synthesis vessel with fritted disc, T-bore PTFE stopcock and vacuum take-off
10 mL, joint: ST/NS 24/40, porosity: medium, GL 14
Synonym(s):
Synthware P140010M, Solid phase peptide synthesis vessel
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware P140010M
capacity
10 mL
10 mL
fitting
GL 14
General description
A connection of 8 mm O.D. is sealed on the inner joint for connection to an inner gas. With a fritted disc of coarse or medium porosity to support resin. Top of the vessel has GL thread and a PTFE lined screw cap. PTFE stopcock has a 2 mm bore with the arm being 8 mm O.D.
Legal Information
Synthware is a trademark of Kemtech America Inc.