Key Documents
SYND201922
Synthware vertical distilling adapter
Cow Receiver Only, joint: ST/NS 19/22
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
Joint
joint: ST/NS19/22
feature
Cow Receiver Only
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D201922
General description
Designed without hose connection to be used with D10 distillation apparatus.
Legal Information
Synthware is a trademark of Kemtech America Inc.