Key Documents
SYNC562280
Synthware coil-style reflux condenser with removable hose connections
joint: ST/NS 14/20, hose size 8 mm
Synonym(s):
Synthware C562280, coil-type condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C562280
hose size
8 mm
General description
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.