Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD10513

Sigma-Aldrich

Azido-dPEG®12-acid

Synonym(s):

Polyethylene glycol, alpha-(2-azidoethyl)-omega-(2-carboxyethoxy)poly(oxy-1,2-ethanediyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H53N3O14
CAS Number:
Molecular Weight:
643.72
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent
reaction type: click chemistry

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

OC(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-])=O

InChI

1S/C27H53N3O14/c28-30-29-2-4-34-6-8-36-10-12-38-14-16-40-18-20-42-22-24-44-26-25-43-23-21-41-19-17-39-15-13-37-11-9-35-7-5-33-3-1-27(31)32/h1-26H2,(H,31,32)

InChI key

DYFODOGACDHUBQ-UHFFFAOYSA-N

Features and Benefits

The azido-dPEG®12 contains an azide function on one end of a single molecular weight dPEG® spacer (46.4 Å) and a reactive group on the other end of the spacer. The dPEG® spacer is hydrophilic and non-immunogenic and improves the water solubility of the target molecule while reducing the immunogenicity and increasing the hydrodynamic volume of the target. The reactive acid functional group may be activated with 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide (EDC) or N,N′-dicyclohexylcarbodiimide (DCC) for conjugation to an amine. The azide group reacts with an alkyne in the well-known click chemistry reaction. The click chemistry reaction proceeds by copper(I) or ruthenium catalysis or in a strain-catalyzed reaction with certain types of alkyne partners.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service