Skip to Content
MilliporeSigma
All Photos(1)

Documents

QBD10406

Sigma-Aldrich

MAL-dPEG®4-(m-dPEG®12)3

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C106H202N6O50
CAS Number:
Molecular Weight:
2360.75
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reactivity: thiol reactive
reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: sulfhydryl reactive

polymer architecture

shape: branched
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCOCCOCCOCCOCCNC(CCN1C(C=CC1=O)=O)=O)NC(COCCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)(COCCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)COCCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O

Features and Benefits

MAL-dPEG®4-(m-dPEG®12)3 is a branched dPEG® reagent which is highly useful for protein and peptide modification to increase hydrodynamic volume and serum half-life and to greatly reduce the immunogenicity of the modified peptide or protein. This dPEG® product consists of a linear "tail" with a reactive group, a tris core to provide branching, and methoxy-terminated dPEG® branches. The numbers following dPEG® are the exact number of repeat units in the linear chain. All of these dPEG® products are hydrophilic, non-immunogenic, single molecular weight compounds. The maleimide functional group reacts rapidly and specifically with free sulfhydryls in aqueous media at pH 6.5-7.5.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service