Skip to Content
MilliporeSigma
All Photos(1)

Documents

P5301

Sigma-Aldrich

Pentafluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC6F5
CAS Number:
Molecular Weight:
168.06
Beilstein:
1911549
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.391 (lit.)

bp

85 °C (lit.)

mp

−48 °C (lit.)

density

1.514 g/mL at 25 °C (lit.)

SMILES string

Fc1cc(F)c(F)c(F)c1F

InChI

1S/C6HF5/c7-2-1-3(8)5(10)6(11)4(2)9/h1H

InChI key

WACNXHCZHTVBJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria A Sannikova et al.
The journal of physical chemistry. A, 123(2), 505-516 (2018-12-20)
Magnetic field-affected reaction yield (MARY) spectroscopy is a spin chemistry technique for detecting short-lived radical ions. Having sensitivity to transient species with lifetimes as short as nanoseconds, MARY spectroscopy usually does not provide detailed information on their magnetic resonance parameters
Leila Hejazi et al.
Rapid communications in mass spectrometry : RCM, 29(1), 91-99 (2014-12-03)
Mass spectrometric identification of compounds in chromatography can be obtained from molecular masses from soft ionization mass spectrometry techniques such as field ionization (FI) and fragmentation patterns from hard ionization techniques such as electron ionization (EI). Simultaneous detection by EI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service