Recommended Products
material
gold/chromium electrode (Au/Cr)
silica coating (SiO2)
description
die layer thickness 300 nm SiO2
electrode layer thickness 60 nm gold
type 1
packaging
pkg of 12 diced devices
manufacturer/tradename
IDE100D
die thickness
525 μm , silicon
electrode size
10 nm , chrome
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The spacing between the fingers on IDE100D is 50um. The fingers vary in thickness between 1um and 5um and in the number of fingers per device, from 9 to 13 fingers. The fingers are 550um long. Each device is about 1mmx1mm.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service