Skip to Content
MilliporeSigma
All Photos(1)

Documents

I6903

Sigma-Aldrich

Iodoacetonitrile

98%

Synonym(s):

Cyanoiodomethane, Cyanomethyl iodide, Iodomethyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2CN
CAS Number:
Molecular Weight:
166.95
Beilstein:
1734628
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.574 (lit.)

bp

182-184 °C/720 mmHg (lit.)

density

2.307 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ICC#N

InChI

1S/C2H2IN/c3-1-2-4/h1H2

InChI key

VODKOOOHHCAWFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service